Learn More
Selleck Chemical LLC Lobeliae Chinensis Extract E3049-5MG
SDP
Supplier:  Selleck Chemical LLC E30495MG
Encompass_Preferred
Lobeliae Chinensis Extract is extracted from Lobeliae Chinensis which is a traditional Chinese herb widely used in the treatment of diabetes and found to have a hypoglycaemic mechanism related to the inhibition of DPP4
Catalog No. 50-000-40706