Learn More
Medchemexpress LLC Lobeline hydrochloride | 134-63-4 | MFCD00082455 | 99.9% | 373.92 g/mol | C22H28ClNO2 | 10 MG

Supplier: Medchemexpress LLC HYB097910MG
Lobeline hydrochloride is a nicotinic receptor agonist used in pharmacology and neuroscience research. It is a brain-penetrant alkaloid that modulates nicotinic acetylcholine receptors and can increase dopamine release; it is supplied for laboratory research in solid and solution formats with supporting safety and technical documentation.
- High purity (>99.8%).
- Molecular weight 373.92 g/mol.
- CAS number 134-63-4.
- Available in multiple pack sizes including 10 MG solid and solution formats.
- Supplied with SDS and product data sheet for handling and safety information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.