Learn More
Medchemexpress LLC Lobendazole | 6306-71-4 | 99.49% | 25 MG

Supplier: Medchemexpress LLC 25MGHYB1313
Lobendazole is a potent anthelmintic that inhibits *Giardia intestinalis* strains. It is for research use only and not sold to patients.
- Potent anthelmintic
- Inhibits Giardia intestinalis strains
- White to off-white solid appearance
- Molecular weight: 205.21
- Formula: C10H11N3O2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.