Learn More
Medchemexpress LLC HY-100432 25mg Medchemexpress, LOC14 CAS:877963-94-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10043225MG
Encompass_Preferred
Medchemexpress, HY-100432 25mg LOC14 CAS:877963-94-5 LOC14 is a potent Protein disulfide isomerase (PDI) inhibitor with EC50 and Kd values of 500 nM and 62 nM, respectively. LOC14 exhibits high stability in mouse liver microsomes and blood plasma, low intrinsic microsome clearance, and low plasma-protein binding.LOC14 inhibits PDIA3 activity, decreases intramolecular disulfide bonds and subsequent oligomerization (maturation) of HA in lung epithelial cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7731