Learn More
Grainger Lockwire Spool 0.0319 Dia 81.5ft
Supplier:  Grainger 16Y059
Encompass_Preferred
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a lock effect when installed in a crisscross pattern to secure bolts, screws, nuts and other threaded assemblies. Its stainless steel construction provides corrosion resistance. With a working temp range of -100F to 500F, this lockwire is suitable for a variety of aerospace, industrial, and automotive applications. The manufacturer part number for this product is 16Y059.
Catalog No. 50-276-6825