Learn More
Grainger Lockwire Spool 0.041 Dia 1105 ft.
Supplier:  Grainger 16Y055
Encompass_Preferred
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a lock effect when installed in a crisscross pattern to secure bolts, nuts, turnbuckles and other threaded assemblies. It conforms to MIL-W-83420 and MS20995 standards. The lockwire is made in the USA.
Catalog No. 50-276-7855