Learn More
Medchemexpress LLC Locust bean gum, from Ceratonia siliqua seeds | 9000-40-2 | MFCD00131257 | >75.0% | Approximately 310 kDa | No fixed formula (polymeric galactomannan); commonly represented as (C6H10O5)n | 1 G
SDP
Catalog No. 5000281385
Encompass_Preferred
Locust bean gum is a natural galactomannan polysaccharide derived from carob (Ceratonia siliqua) seeds. It is used as a thickening, stabilizing, and gelling agent in food, pharmaceutical, cosmetic, and laboratory applications due to its high water retention and compatibility with other hydrocolloids.
  • High-viscosity powder with typical viscosity 5,000-6,500 mPa·s.
  • Polysaccharide content >75%.
  • Off-white to light-yellow solid powder.
  • Forms gels and stabilizes emulsions; synergistic with other hydrocolloids.
  • Suitable as a dietary fiber and texturizer in formulations.
  • Polymeric galactomannan commonly represented as (C6H10O5)n; variable molecular weight.
Catalog No. 50-002-81385 Supplier Medchemexpress LLC Supplier No. HYW2501461G
Locust bean gum is a natural galactomannan polysaccharide derived from carob (Ceratonia siliqua) seeds. It is used as a thickening, stabilizing, and gelling agent in food, pharmaceutical, cosmetic, and laboratory applications due to its high water retention and compatibility with other hydrocolloids.
  • High-viscosity powder with typical viscosity 5,000-6,500 mPa·s.
  • Polysaccharide content >75%.
  • Off-white to light-yellow solid powder.
  • Forms gels and stabilizes emulsions; synergistic with other hydrocolloids.
  • Suitable as a dietary fiber and texturizer in formulations.
  • Polymeric galactomannan commonly represented as (C6H10O5)n; variable molecular weight.