Learn More
Medchemexpress LLC HY-12390 10mg , Lofepramine CAS:23047-25-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1239010MG
Encompass_Preferred
HY-12390 10mg Lofepramine CAS: 23047-25-8 Lofepramine (Lopramine) is a potent tricyclic antidepressant and is extensively metabolised to Desipramine. The antidepressant activity of Lofepramine stems from the facilitation of noradrenergic neurotransmission by uptake inhibition. Lofepramine may also potentiate serotoninergic neurotransmission by inhibition of the neuronal uptake of serotonin and the enzyme tryptophan pyrrolase. Lofepramine has significant anxiolytic efficacy in addition to its antidepressant properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9994