Learn More
Cayman Chemical LofexIdInhydrochlorIde 1g
SDP
Supplier:  Cayman Chemical 159891 G
Encompass_Preferred
An α2-adrenergic receptor agonist (Kd = 7.6 nM for rat cerebral cortex membranes) that has transient antihypertensive effects; used in managing opioid withdrawal symptoms during detoxification from heroin and methadone
Catalog No. 50-296-8536