Learn More
Medchemexpress LLC Loganic acid | 22255-40-9 | 376.36 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0513
Encompass_Preferred
Loganic acid is an iridoid isolated from cornelian cherry fruits. It inhibits the NF-κB signaling pathway, activates the Nrf2 signaling pathway, and exhibits anti-inflammatory activity. It can modulate diet-induced atherosclerosis and redox status, possesses strong free radical scavenging activity, and a remarkable cyto-protective effect against heavy metal mediated toxicity. Loganic acid is orally active.
  • Isolated from cornelian cherry fruits.
  • Inhibits NF-κB signaling pathway.
  • Activates Nrf2 signaling pathway.
  • Exhibits anti-inflammatory activity.
  • Modulates diet-induced atherosclerosis and redox status.
  • Strong free radical scavenging activity.
  • Remarkable cyto-protective effect against heavy metal mediated toxicity.
  • Orally active.
Catalog No. 50-003-89490