Learn More
Selleck Chemical LLC Loganic acid S9191-1MG
SDP
Supplier:  Selleck Chemical LLC S91911MG
Encompass_Preferred
Loganic acid (Loganate) is a naturally occurring iridoid monoterpeneglusoside involved in the biosynthesis of indole alkaloids in Vinca rosea
Catalog No. 50-000-25169