Learn More
Indofine Chemical Loganin With Hplc, 10 Mg
SDP
Supplier:  Indofine Chemical 020231S 10 MG
Encompass_Preferred
LOGANIN with HPLC 10 MG, Chromatography Standards, 7-Hydroxy-6-desoxyverbenalin, C17H26O10, MW: 390.39, 99% by HPLC, 18524-94-2, MFCD00075645
Catalog No. 50-908-847