Learn More
Medchemexpress LLC LolCDE-IN-2 | 2748443-01-6 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13084010MM/1ML
Encompass_Preferred
LolCDE-IN-2 is a potent inhibitor of the Lol protein (LolCDE), demonstrating antibacterial activity. It effectively inhibits E. coli MG1655 and increases the susceptibility of P. aeruginosa ΔlolCDE::CTX-lolCDE E. coli strain under specific conditions.
  • Potent Lol protein (LolCDE) inhibitor.
  • Exhibits antibacterial activity.
  • Inhibits E. coli MG1655 with an MIC of 2 μg/ml.
  • Increases susceptibility of P. aeruginosa ΔlolCDE::CTX-lolCDE E. coli strain under specific conditions.
Catalog No. 50-003-40738