Learn More
Medchemexpress LLC Lomefloxacin-d5 hydrochloride | 98079-52-8 | 99.88% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0455S5MG
Encompass_Preferred
Lomefloxacin-d5 (hydrochloride) is the deuterium-labeled Lomefloxacin hydrochloride. Lomefloxacin (SC47111A) hydrochloride is a broad-spectrum quinolone antibiotic with antimicrobial activity. It is used for the research of respiratory tract infections, genitourinary infections, gastrointestinal infections, and ENT infections. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, involving the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is used as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06058