Learn More
Medchemexpress LLC Lomefloxacin (hydrochloride) | 98079-52-8 | 100.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB045510G
Encompass_Preferred
Lomefloxacin hydrochloride is an orally active difluoroquinolone antibiotic that inhibits bacterial topoisomerase II, preventing DNA supercoiling and replication. It induces ROS production and apoptosis, exhibiting broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria, and anticancer effects against melanoma.
  • Inhibits bacterial topoisomerase II
  • Prevents DNA supercoiling and replication
  • Induces ROS production and apoptosis
  • Broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria
  • Exhibits anticancer effects against melanoma
  • Used in the study of systemic bacterial infections, skin, and melanoma
Catalog No. 50-004-07762