Learn More
Medchemexpress LLC Lomefloxacin (hydrochloride) | 98079-52-8 | 100.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0455500MG
Encompass_Preferred
Lomefloxacin hydrochloride is an orally active difluoroquinolone antibiotic. It functions by preventing DNA supercoiling and replication through the inhibition of bacterial topoisomerase II. This compound is known to induce ROS production and apoptosis, demonstrating broad-spectrum bactericidal activity against both Gram-positive and Gram-negative bacteria. It also exhibits anticancer effects, particularly against melanoma.
  • Orally active difluoroquinolone antibiotic
  • Inhibits bacterial topoisomerase II, preventing DNA supercoiling and replication
  • Induces ROS production and apoptosis
  • Provides broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria
  • Exhibits anticancer effects against melanoma
  • Useful for studying systemic bacterial infections, skin, and melanoma
  • Rapidly reduces viable cell counts of common bacteria
Catalog No. 50-004-06059