Learn More
TARGETMOL CHEMICALS INC LOMITAPIDE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T626710MG
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lomitapide (AEGR-733) is a small molecule inhibitor of microsomal triglyceride transfer protein (MTP) an enzyme located in the lumen of the endoplasmic reticulum responsible for absorbing dietary lipids and transferring triglycerides onto apolipoprotein B (apo-B) in the assembly of very-low-density lipoprotein. purity: 99%
Catalog No. 50-257-9466