Learn More
Medchemexpress LLC Lomitapide | 182431-12-5 | MFCD16620494 | 99.6% | 693.72 | C39H37F6N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146675MG
Encompass_Preferred
Lomitapide is a research-grade inhibitor of microsomal triglyceride-transfer protein (MTP) provided as a high-purity solid for laboratory and analytical applications. It exhibits potent in vitro activity against MTP and is used in biochemical and cellular studies for mechanism-of-action and screening work.
  • High purity solid suitable for analytical and biochemical assays.
  • Potent MTP inhibition with IC50 ≈ 8 nM for sensitive activity profiling.
  • Supplied in milligram-scale quantities for screening and validation workflows.
  • Well characterized with molecular formula and molecular weight for compound identification.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-22141