Learn More
Medchemexpress LLC Lomitapide mesylate | 202914-84-9 | MFCD19443682 | 99.6% | 789.83 g/mol | C40H41F6N3O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146685MG
Encompass_Preferred
Lomitapide mesylate is the mesylate salt form of lomitapide, a potent inhibitor of microsomal triglyceride transfer protein (MTP). It is supplied as a high-purity analytical standard for in vitro and biochemical research, with a reported IC50 of 8 nM. The product is intended for analytical and research applications where a stable, well-characterized MTP inhibitor is required.
  • Potent MTP inhibitor with reported IC50 of 8 nM.
  • High purity suitable for analytical and in vitro research.
  • Provided as a stable solid, convenient for formulation and storage.
  • Molecular formula C40H41F6N3O5S and molecular weight ≈ 789.8 g/mol.
  • Available in small-scale quantities for analytical use.
Catalog No. 50-002-22642