Learn More
Apexbio Technology LLC Lonafarnib 193275-84-2 100mg
SDP
Supplier:  Apexbio Technology LLC A4379100
Encompass_Preferred
Lonafarnib (CAS 193275-84-2) also known as SCH66336 or Sarasar is a tricyclic orally bioavailable inhibitor of farnesyltransferase (FTase) functioning without peptide or sulfhydryl groups FTase mediates post-translational farnesylation of Ras proteins a modification required for their membrane localization and activation of downstream signaling pathways involved in cellular growth proliferation and survival By inhibiting this enzymatic activity Lonafarnib disrupts Ras translocation to the plasma membrane limiting Ras-mediated signaling Currently Lonafarnib is widely evaluated in preclinical and clinical investigations particularly in oncology contexts
Catalog No. 50-000-61870