Learn More
Apexbio Technology LLC Lonafarnib, 10mg. Cas: 193275-84-2 MFCD: MFCD06795138. Ftase inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A437910
Encompass_Preferred
Lonafarnib (SCH66336, Sarasar) is an potent, selective, orally, bioavailable tricyclic nonpeptidyl nonsulfhydry inhibitor of farnesyltransferase (FTase).[1] It is a small molecular with the formula of C27H31Br2ClN4O2 and molecular weight of 638.82. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4935