Learn More
Medchemexpress LLC HY-15136 5mg Medchemexpress, Lonafarnib CAS:193275-84-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY151365MG
Encompass_Preferred
Medchemexpress, HY-15136 5mg Lonafarnib CAS:193275-84-2 Lonafarnib is an orally bioavailable farnesyl protein transferase (FPTase) inhibitor for H-ras, K-ras and N-ras with IC50 of 1.9 nM, 5.2 nM and 2.8 nM, respectively. Lonafarnib has anti-hepatitis delta virus (HDV) activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0585
 Restricted Item