Learn More
Neta Scientific Agilent Deuterium lamp 8-pin w. RFID for DAD
SDP
Supplier:  Neta Scientific 51900917
Encompass_Preferred
This item is manufactured by Agilent
Agilent InfinityLab long-life HiS deuterium lamp, 8 pin, with RFID tag, for diode array detectors with Max-Light cartridge cell (G4212A/B, G7117A/B/C/D) and G7100 CE. - Repair Parts
UNSPSC Code  39101600
- 58
Catalog No. 50-011-0295