Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump Pump head YZ35-13 PSU Housing, Flow Rate: 600-11000ml/min, High-Quality Rollers, Easy Tubing Change, Automatic Retention, High Temperature Resistance
Supplier:  LONGER PUMP 3/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump Pump head YZ35-13Applications:peristaltic pump head suitable for all kind of industry and labProduct Instruction:Change tubing easily and rapidly; Automatic tubing retention; Suitable for several tubing sizes;The rollers adopt high quality materials and has good wearing property; The housing of the pump head has perfect rigidity and structure property, and its material is PSU which has stable performance and high temperature resistance (150 °C).
Catalog No. NC2956847