Learn More
Medchemexpress LLC Lonicerin | 25694-72-8 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN4136
Encompass_Preferred
Lonicerin, also known as Veronicastroside, is a flavonoid with a range of biological activities. It is known to inhibit xanthine oxidase, alginate secretion protein (AlgE), *P. aeruginosa*, and *C. albicans*.
  • Inhibits xanthine oxidase
  • Acts against alginate secretion protein (AlgE)
  • Demonstrates antibacterial activity against *P. aeruginosa*
  • Exhibits antifungal properties against *C. albicans*
  • Provides anti-inflammatory effects
  • Has anti-arthritic properties
  • Functions as an antioxidant
  • Offers neuroprotective benefits
  • Reduces biofilm formation
  • Suppresses T cell proliferation
  • Inhibits nitric oxide production
Catalog No. 50-004-21381