Learn More
Medchemexpress LLC Lonicerin (Veronicastroside) | 25694-72-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN41365MG
Encompass_Preferred
Lonicerin (Veronicastroside) is a flavonoid that exhibits a range of biological activities. It functions as an inhibitor of xanthine oxidase and alginate secretion protein (AlgE). Additionally, Lonicerin demonstrates anti-inflammatory, anti-arthritic, antioxidant, antifungal, anti-bacterial, and neuroprotective properties.
  • Inhibits xanthine oxidase, alginate secretion protein (AlgE), P. aeruginosa, and C. albicans.
  • Possesses anti-inflammatory, anti-arthritic, and neuroprotective properties.
  • Acts as an antioxidant, scavenging DPPH, hydroxyl, and superoxide radicals.
  • Reduces alginate secretion and biofilm formation.
  • Suppresses T cell proliferation and inhibits nitric oxide production from macrophages.
  • Reduces paw edema in a mouse model of C. albicans arthritis.
Catalog No. 50-004-20394