Learn More
Grainger Loop Round 3/8 Dia White PK1890
Supplier:  Grainger 16U769
Encompass_Preferred
The Grainger website features hook and loop cable ties for wire and cable management. These reusable ties are available in various sizes, lengths, and tensile strengths to securely bundle wires and cables. They feature a hook and loop fastening system for easy installation, removal, and repositioning. Offered in black and other colors, these ties can help organize and route cables in telecom, electrical, industrial, and IT applications. Grainger provides a wide selection of reputable brands like 3M, Panduit, and Velcro.
Catalog No. 50-276-7232