Learn More
Selleck Chemical LLC Loperamide HCl S2480-50mg
SDP
Supplier:  Selleck Chemical LLC S248050MG
Encompass_Preferred
Loperamide HCl (ADL 2-1294 R-18553 hydrochloride) is a selective -opioid receptor agonist opioid with Ki of 3 3 nM 15-fold and 350-fold selective over the subtype and the subtype of the opioid receptor used against diarrhea resulting from gastroenteritis or inflammatory bowel disease
Catalog No. 50-311-1664