Learn More
Selleck Chemical LLC Loratadine S1358-50mg
SDP
Supplier:  Selleck Chemical LLC S135850MG
Encompass_Preferred
Loratadine is a histamine H1 receptor antagonist used to treat allergies Also acts as a selective inhibitor of B(0)AT2 with IC50 of 4 M
Catalog No. 50-311-0704