Learn More
Medchemexpress LLC Loratadine | 79794-75-5 | 100.0% | 382.88 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17043100MG
Encompass_Preferred
Loratadine (SCH-29851) is a selective inverse peripheral histamine H1-receptor agonist that exhibits anti-dengue-virus (DENV) activity. It is capable of inhibiting the immunologic release of inflammatory mediators and is provided as a white to off-white solid.
  • Selective inverse peripheral histamine H1-receptor agonist
  • Exhibits anti-dengue-virus (DENV) activity
  • Inhibits immunologic release of inflammatory mediators
  • Tested and complies with given specifications
Catalog No. 50-003-84675