Learn More
Medchemexpress LLC Loratadine-d5 | 1398065-63-8 | 99.96% | 387.91 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17043S11MG
Encompass_Preferred
Loratadine-d5 is a deuterium labeled variant of Loratadine, a selective inverse peripheral histamine H1-receptor agonist. This compound shows anti-dengue-virus (DENV) activity and can inhibit the immunologic release of inflammatory mediators, with an IC50 of >32 μM. It is intended for research use only, serving as a valuable tool in drug development and quantitative analysis due to its stable heavy isotope labeling.
  • Can be used as a tracer in research studies.
  • Serves as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Exhibits anti-dengue-virus (DENV) activity.
  • Inhibits the immunologic release of inflammatory mediators.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-84889