Learn More
Medchemexpress LLC Loratadine (Standard) | 79794-75-5 | 99.9% | 382.88 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1704350MG
Encompass_Preferred
Loratadine (Standard) is the analytical standard for Loratadine, intended for research and analytical applications. It acts as a selective inverse peripheral histamine H1-receptor agonist with an IC50 of >32 μM. This compound exhibits anti-dengue-virus (DENV) activity and can inhibit the immunologic release of inflammatory mediators. It is categorized as an analytical standard grade and is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard of Loratadine
  • Intended for research and analytical applications
  • Selective inverse peripheral histamine H1-receptor agonist
  • Exhibits anti-dengue-virus (DENV) activity
  • Can inhibit immunologic release of inflammatory mediators
  • Commonly used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-85875