Learn More
Medchemexpress LLC Loratadine (Standard) | 79794-75-5 | MFCD00672869 | 99.9% | 382.88 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17043R
Encompass_Preferred
Loratadine (Standard) is an analytical standard intended for research and analytical applications. It acts as a selective inverse peripheral histamine H1-receptor agonist and has anti-dengue-virus (DENV) activity. This compound can also inhibit the immunologic release of inflammatory mediators.
  • Selective inverse peripheral histamine H1-receptor agonist
  • Anti-dengue-virus (DENV) activity
  • Inhibits immunologic release of inflammatory mediators
  • Analytical standard for research and analytical applications
Catalog No. 50-003-84201