Learn More
U.S. Pharmacopeia Lorazepam Related Compound B, 2958-36-3, MFCD00007840, 25 mg
Supplier:  U.S. Pharmacopeia 1370338
Encompass
Linear Formula: H2NC6H3(Cl)COC6H4Cl, Molecular Weight: 266.12, mp: 87-89 °C, Synonym: 2-Amino-2′,5-dichlorobenzophenone.
Catalog No. NC1087017