Learn More
Medchemexpress LLC HY-105272 5mg Medchemexpress, Loreclezole CAS:117857-45-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1052725MG
Encompass_Preferred
Medchemexpress, HY-105272 5mg Loreclezole CAS:117857-45-1 Loreclezole, an antiepileptic compound, is a selective GABA A receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2260