Learn More
Enterprise Technology Solutions Lorell Barrier,w/Cutout,f/Social Distance,Acrylic,36"x7"x30",CL
SDP
Supplier:  Enterprise Technology Solutions LLR55685
Encompass_Preferred
Social Distancing Barrier with cutout provides added protection from respiratory droplets for both the customer and employee. They can interact and exchange communication with a dependable barrier between them. Cutout allows them to exchange papers and other small objects. Barrier is freestanding on a triangular base so it is easy to move and reposition in a variety of countertop settings. The 1/4" thick acrylic offers clarity and durability.
Catalog No. 50-007-91403