Learn More
Enterprise Technology Solutions Lorell Lateral File, 3-Drawer, 36"x18-5/8"x40-1/4", Putty
SDP
Supplier:  Enterprise Technology Solutions LLR88027
Encompass_Preferred
36" wide lateral file features three versatile drawers with hanging file rails for side-to-side filing in letter-size, legal-size or A4-size. Each drawer has a magnetic label holder for easy identification where you need it. Design also features a core-removable lock for security, dual locking bars, full-width pull, ball-bearing slide suspension for smooth opening and closing, reinforced base, and four adjustable leveling guides for stabilization. Interlock system prohibits the extension of more than one drawer at a time for your safety. Counterweight prevents tipping. Lateral file meets or exceeds applicable ANSI/BIFMA standards.
Catalog No. 50-007-91700