Learn More
Enterprise Technology Solutions Lorell Shelving, 5-Shelf, Steel, 2300 lb Cap, 48"x18"x72", Black
SDP
Supplier:  Enterprise Technology Solutions LLR59698
Encompass_Preferred
Iron Horse Riveted Steel Shelving offers a boltless design for simple assembly. It holds up to 2,300 lb. of evenly distributed weight on its particleboard shelving. The five shelves are supported by a black steel frame with a powder-coat finish for durability. Shelving assembles vertically or horizontally.
Catalog No. 50-007-91250