Learn More
Enterprise Technology Solutions Lorell Shelving,Wire,4 Shelves/4 Posts,36"x18"x72",CE
SDP
Supplier:  Enterprise Technology Solutions LLR84187
Encompass_Preferred
Industrial wire shelving is designed to provide the essentials for industrial wire shelving so you can organize your storage areas. Kit includes four shelves and four posts. Easy collar-and-sleeve assembly requires no tools. Each shelf adjusts in 1" increments. Shelving is made of steel with a chrome powder-coat finish, 10-gauge shelf surface, 3-gauge frame and 6-gauge wave pattern. Weight capacity is 1,000 lb. evenly distributed.
Catalog No. 50-007-91626