Learn More
Enterprise Technology Solutions Lorell Shelving, Wire, Boltless, 2300 lb. Cap, 36"x18"x60", Black
SDP
Supplier:  Enterprise Technology Solutions LLR99928
Encompass_Preferred
Wire Deck Shelving features a boltless design for easy assembly and professional appearance. It holds a total capacity of 2300 lb. evenly distributed on its four shelves. Each shelf supports 460 lb. Easy-to-clean shelves are supported by a heavy-duty steel frame with a rust-resistant, powder-coat finish for durability. Shelving assembles vertically or horizontally with a rubber mallet.
Catalog No. 50-007-91295