Learn More
Cayman Chemical LorglumIdesodIum salt 50mg
SDP
Supplier:  Cayman Chemical 1755550 MG
Encompass_Preferred
A nonpeptidic antagonist of CCK1 (IC50 = 50 nM) that is 60-fold less effective at CCK2 (IC50 = 3 µM); blocks CCK-mediated gut muscle contraction, pancreatic growth, and pancreatic secretion; commonly used to study the roles of CCK1 in tissues and in animals
Catalog No. 50-296-6881