Learn More
Cayman Chemical LornoxIcam-d4 1mg
SDP
Supplier:  Cayman Chemical 332751 MG
Encompass_Preferred
An internal standard for the quantification of lornoxicam by GC- or LC-MS
Catalog No. 50-294-6289