Learn More
Medchemexpress LLC Lorundrostat | 1820940-17-7 | 99.4% | 451.56 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147277100MG
Encompass_Preferred
Lorundrostat (MT-4129) is an orally active aldosterone synthase inhibitor that can be used in studies of high blood pressure.
  • Orally active
  • Aldosterone synthase inhibitor
  • Used in studies of high blood pressure
Catalog No. 50-003-62059