Learn More
Medchemexpress LLC Lorundrostat | 1820940-17-7 | 99.4% | C24H33N7O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14727710MG
Encompass_Preferred
Lorundrostat is an orally active, selective aldosterone synthase (CYP11B2) inhibitor used in research on blood pressure regulation and mineralocorticoid-related pathophysiology. It reduces aldosterone production and has been evaluated in preclinical and clinical studies for antihypertensive effects. The compound is supplied as a light yellow solid with high purity and characterized chemical properties suitable for in vitro and in vivo pharmacology studies.
  • Selective inhibition of aldosterone synthase (CYP11B2) for targeted aldosterone reduction.
  • Oral activity demonstrated in preclinical and clinical studies relevant to hypertension.
  • High purity suitable for pharmacology and toxicology assays.
  • Soluble in DMSO and compatible with common in vivo formulation vehicles.
  • Stable under recommended storage conditions for reliable long-term use.
Catalog No. 50-002-58661