Learn More
Selleck Chemical LLC Losartan S5067-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S506710MM/1ML
Encompass_Preferred
Losartan(DuP-753) is a selective orally administered nonpeptide blocker of angiotensin II type 1 (AT1) receptor used to treat high blood pressure diabetic kidney disease heart failure and left ventricular enlargement
Catalog No. 50-311-0166