Learn More
Selleck Chemical LLC Losartan Carboxylic Acid
SDP
Supplier:  Selleck Chemical LLC S59805MG
Encompass_Preferred
Losartan Carboxylic Acid (E-3174 EXP-3174) an active carboxylic acid metabolite of Losartan is a non-peptide angiotensin II (AT1) receptor antagonist Losartan Carboxylic Acid inhibits the specific binding of [125I]-angiotensin II to VSMC with IC50 of 1 1 nM
Catalog No. 50-000-04095