Learn More
Medchemexpress LLC Losartan-d4 | 1030937-27-9 | 99.9% | 426.94 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17512S1MG
Encompass_Preferred
Losartan-d4 is the deuterium-labeled Losartan, an angiotensin II receptor antagonist that competes with the binding of angiotensin II to AT1 receptors with an IC50 of 20 nM.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86286