Learn More
Cayman Chemical Losartan-d4 1mg
SDP
Supplier:  Cayman Chemical 225671 MG
Encompass_Preferred
An internal standard for the quantification of losartan (potassium salt) by GC- or LC-MS
Catalog No. 50-296-4293