Learn More
Medchemexpress LLC Losartan (potassium) | 124750-99-8 | 99.98% | 461.00 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY17512A10MM/1ML
Encompass_Preferred
Losartan potassium (DuP-753 potassium) is an angiotensin II receptor type 1 (AT1) antagonist, competing with the binding of angiotensin II to AT1 with an IC50 of 20 nM.
  • Targets angiotensin receptor
  • Used in research for cardiovascular disease, endocrinology, and cancer
  • Functions as an antagonist of angiotensin II receptor type 1 (AT1)
Catalog No. 50-003-86401