Learn More
Selleck Chemical LLC Losartan Potassium (DuP 753) 50mg 124750-99-8 MK 954
SDP
Supplier:  Selleck Chemical LLC S135950MG
Encompass_Preferred
Losartan Potassium (DuP 753, MK 954) is an angiotensin II receptor antagonist, competes with the binding of angiotensin II to AT1 receptors with IC50 of 20 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1333431